Product Details

SNP ID
rs201591425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104535892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCATAAAACAGAGAAATGAGCT[C/T]GTCTAATGCTTGCAATTTTTCTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13C3 PubMed Links

Gene Details

Gene
OR13C3
Gene Name
olfactory receptor family 13 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001961.1 922 Missense Mutation AAG,GAG K308E NP_001001961.1
Gene
OR13C4
Gene Name
olfactory receptor family 13 subfamily C member 4
There are no transcripts associated with this gene.

View Full Product Details