Product Details

SNP ID
rs202004319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:12776018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAACCAAGCTTCACCTCCTCAG[A/G]CAAGAGATGGTGAGTGTGGTGCGCC
Phenotype
MIM: 616130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LURAP1L PubMed Links

Gene Details

Gene
LURAP1L
Gene Name
leucine rich adaptor protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203403.1 1006 Silent Mutation AGA,AGG R101R NP_981948.1
XM_005251443.2 1006 Silent Mutation AGA,AGG R101R XP_005251500.1
Gene
LURAP1L-AS1
Gene Name
LURAP1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details