Product Details

SNP ID
hCV192183414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48835425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGGACTGGGGTCGCACCTTTAC[T/C]GGCCGCGCGCAGAGCGCGGGGGGCG
Phenotype
MIM: 300437 MIM: 300399
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ERAS PubMed Links

Gene Details

Gene
ERAS
Gene Name
ES cell expressed Ras
There are no transcripts associated with this gene.

Gene
PCSK1N
Gene Name
proprotein convertase subtilisin/kexin type 1 inhibitor
There are no transcripts associated with this gene.

View Full Product Details