Product Details

SNP ID
rs199597806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152134548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAAGCTACCTGTAGCGCTAGAA[C/G]TTGCACTGGCCATGGCAGTAGTATC
Phenotype
MIM: 300343
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAGEA10 PubMed Links

Gene Details

Gene
MAGEA10
Gene Name
MAGE family member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011543.2 1506 Missense Mutation ACT,AGT T358S NP_001011543.2
NM_001251828.1 1506 Missense Mutation ACT,AGT T358S NP_001238757.1
NM_021048.4 1506 Missense Mutation ACT,AGT T358S NP_066386.2
Gene
MAGEA10-MAGEA5
Gene Name
MAGEA10-MAGEA5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204811.2 1506 Intron NP_001191740.1

View Full Product Details