Product Details

SNP ID
rs199861985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154460409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGGGCCCGTCGAGGACAACGCT[C/T]GCTGCTACCCGCCCCCCAGCATGCG
Phenotype
MIM: 300453 MIM: 300022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM50A PubMed Links

Gene Details

Gene
FAM50A
Gene Name
family with sequence similarity 50 member A
There are no transcripts associated with this gene.

Gene
PLXNA3
Gene Name
plexin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017514.4 401 Missense Mutation CGC,TGC R76C NP_059984.3
XM_005274705.4 401 Missense Mutation CGC,TGC R76C XP_005274762.2
XM_005274706.4 401 Intron XP_005274763.2
XM_006724829.3 401 Missense Mutation CGC,TGC R76C XP_006724892.1
XM_011531183.2 401 Intron XP_011529485.1

View Full Product Details