Product Details

SNP ID
rs199576118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132078536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTGTCCACATAAAAAAAGACC[C/T]GGGGAGGCATAATACCAACCTGCTG
Phenotype
MIM: 300628 MIM: 300547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRMD7 PubMed Links

Gene Details

Gene
FRMD7
Gene Name
FERM domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306193.1 1621 Missense Mutation CAG,CGG Q479R NP_001293122.1
NM_194277.2 1621 Missense Mutation CAG,CGG Q494R NP_919253.1
XM_017029947.1 1621 Missense Mutation CAG,CGG Q478R XP_016885436.1
XM_017029948.1 1621 Missense Mutation CAG,CGG Q409R XP_016885437.1
XM_017029949.1 1621 Missense Mutation CAG,CGG Q336R XP_016885438.1
Gene
STK26
Gene Name
serine/threonine protein kinase 26
There are no transcripts associated with this gene.

View Full Product Details