Product Details

SNP ID
rs200304646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136488201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCATCAAGCTCTGTGGTTTCTAA[C/T]GATAACACAAATAAAGGATGGAGCG
Phenotype
MIM: 300107 MIM: 300346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRS3 PubMed Links

Gene Details

Gene
BRS3
Gene Name
bombesin receptor subtype 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001727.1 236 Silent Mutation AAC,AAT N29N NP_001718.1
Gene
HTATSF1
Gene Name
HIV-1 Tat specific factor 1
There are no transcripts associated with this gene.

View Full Product Details