Product Details

SNP ID
rs199745174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:115190042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTGCAGCCACGGCGCTGGGCC[A/G]GCCCACCCCACAAGAGGGCTGTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRCH2 PubMed Links

Gene Details

Gene
LRCH2
Gene Name
leucine rich repeats and calponin homology domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243963.1 643 Intron NP_001230892.1
NM_020871.3 643 Intron NP_065922.3
XM_006724724.3 643 Intron XP_006724787.2
XM_017029696.1 643 Intron XP_016885185.1
XM_017029697.1 643 Intron XP_016885186.1
Gene
RBMXL3
Gene Name
RNA binding motif protein, X-linked like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145346.1 643 Missense Mutation AGC,GGC S201G NP_001138818.1

View Full Product Details