Product Details

SNP ID
rs200195310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154349695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCCCCAATGTGGTAGGGGCC[A/G]CCGTACTTGATGGAGATGAGGTAGC
Phenotype
MIM: 300017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLNA PubMed Links

Gene Details

Gene
FLNA
Gene Name
filamin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110556.1 7552 Silent Mutation GGC,GGT G2502G NP_001104026.1
NM_001456.3 7552 Silent Mutation GGC,GGT G2494G NP_001447.2
XM_011531127.1 7552 Silent Mutation GGC,GGT G2470G XP_011529429.1
XM_011531128.1 7552 Silent Mutation GGC,GGT G2462G XP_011529430.1
XM_011531129.1 7552 Silent Mutation GGC,GGT G2444G XP_011529431.1
XM_011531130.1 7552 Silent Mutation GGC,GGT G2436G XP_011529432.1
XM_011531131.1 7552 Silent Mutation GGC,GGT G2435G XP_011529433.1
Gene
LOC105373386
Gene Name
uncharacterized LOC105373386
There are no transcripts associated with this gene.

View Full Product Details