Product Details

SNP ID
rs199854559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106612085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTACGCAGTTATCCAACAATGT[C/G]TGGTGAGTCAGGACAGCCTGAGGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CXorf57 PubMed Links

Gene Details

Gene
CXorf57
Gene Name
chromosome X open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184782.1 156 Missense Mutation TCT,TGT S2C NP_001171711.1
NM_018015.5 156 Missense Mutation TCT,TGT S2C NP_060485.4

View Full Product Details