Product Details
- SNP ID
-
rs201210021
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.Y:13326339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGCAGCTATTAGACAAGCCATTT[C/T]TACCTGGATTCCTTTAAAAAGAAGC
- Phenotype
-
MIM: 400009
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UTY
PubMed Links
Gene Details
- Gene
- UTY
- Gene Name
- ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258249.1 |
3041 |
Missense Mutation |
AAA,AGA |
K949R |
NP_001245178.1 |
NM_001258250.1 |
3041 |
Missense Mutation |
AAA,AGA |
K712R |
NP_001245179.1 |
NM_001258251.1 |
3041 |
Missense Mutation |
AAA,AGA |
K868R |
NP_001245180.1 |
NM_001258252.1 |
3041 |
Missense Mutation |
AAA,AGA |
K897R |
NP_001245181.1 |
NM_001258253.1 |
3041 |
Missense Mutation |
AAA,AGA |
K868R |
NP_001245182.1 |
NM_001258254.1 |
3041 |
Missense Mutation |
AAA,AGA |
K773R |
NP_001245183.1 |
NM_001258255.1 |
3041 |
Missense Mutation |
AAA,AGA |
K773R |
NP_001245184.1 |
NM_001258256.1 |
3041 |
Missense Mutation |
AAA,AGA |
K897R |
NP_001245185.1 |
NM_001258257.1 |
3041 |
Missense Mutation |
AAA,AGA |
K728R |
NP_001245186.1 |
NM_001258258.1 |
3041 |
Missense Mutation |
AAA,AGA |
K904R |
NP_001245187.1 |
NM_001258259.1 |
3041 |
Missense Mutation |
AAA,AGA |
K836R |
NP_001245188.1 |
NM_001258260.1 |
3041 |
Missense Mutation |
AAA,AGA |
K882R |
NP_001245189.1 |
NM_001258261.1 |
3041 |
Missense Mutation |
AAA,AGA |
K894R |
NP_001245190.1 |
NM_001258262.1 |
3041 |
Missense Mutation |
AAA,AGA |
K825R |
NP_001245191.1 |
NM_001258263.1 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
NP_001245192.1 |
NM_001258264.1 |
3041 |
Missense Mutation |
AAA,AGA |
K881R |
NP_001245193.1 |
NM_001258265.1 |
3041 |
Missense Mutation |
AAA,AGA |
K769R |
NP_001245194.1 |
NM_001258266.1 |
3041 |
Missense Mutation |
AAA,AGA |
K897R |
NP_001245195.1 |
NM_001258267.1 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
NP_001245196.1 |
NM_001258268.1 |
3041 |
Missense Mutation |
AAA,AGA |
K773R |
NP_001245197.1 |
NM_001258269.1 |
3041 |
Missense Mutation |
AAA,AGA |
K803R |
NP_001245198.1 |
NM_001258270.1 |
3041 |
Missense Mutation |
AAA,AGA |
K728R |
NP_001245199.1 |
NM_007125.4 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
NP_009056.3 |
NM_182659.1 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
NP_872600.1 |
NM_182660.1 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
NP_872601.1 |
XM_005262518.3 |
3041 |
Missense Mutation |
AAA,AGA |
K897R |
XP_005262575.1 |
XM_006724875.3 |
3041 |
Missense Mutation |
AAA,AGA |
K949R |
XP_006724938.1 |
XM_011531441.2 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_011529743.1 |
XM_011531442.2 |
3041 |
Missense Mutation |
AAA,AGA |
K949R |
XP_011529744.1 |
XM_011531443.2 |
3041 |
Missense Mutation |
AAA,AGA |
K943R |
XP_011529745.1 |
XM_011531445.2 |
3041 |
Missense Mutation |
AAA,AGA |
K920R |
XP_011529747.1 |
XM_011531446.2 |
3041 |
Missense Mutation |
AAA,AGA |
K913R |
XP_011529748.1 |
XM_011531447.2 |
3041 |
Missense Mutation |
AAA,AGA |
K904R |
XP_011529749.1 |
XM_011531448.2 |
3041 |
Missense Mutation |
AAA,AGA |
K898R |
XP_011529750.1 |
XM_011531449.2 |
3041 |
Missense Mutation |
AAA,AGA |
K897R |
XP_011529751.1 |
XM_011531450.2 |
3041 |
Missense Mutation |
AAA,AGA |
K882R |
XP_011529752.1 |
XM_011531451.2 |
3041 |
Missense Mutation |
AAA,AGA |
K868R |
XP_011529753.1 |
XM_011531452.2 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
XP_011529754.1 |
XM_011531453.2 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_011529755.1 |
XM_011531454.2 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_011529756.1 |
XM_011531455.2 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_011529757.1 |
XM_011531457.2 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_011529759.1 |
XM_011531458.2 |
3041 |
Missense Mutation |
AAA,AGA |
K927R |
XP_011529760.1 |
XM_011531459.2 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_011529761.1 |
XM_011531460.2 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_011529762.1 |
XM_011531461.2 |
3041 |
Missense Mutation |
AAA,AGA |
K949R |
XP_011529763.1 |
XM_011531462.2 |
3041 |
Missense Mutation |
AAA,AGA |
K927R |
XP_011529764.1 |
XM_011531463.2 |
3041 |
Missense Mutation |
AAA,AGA |
K913R |
XP_011529765.1 |
XM_011531464.2 |
3041 |
Missense Mutation |
AAA,AGA |
K897R |
XP_011529766.1 |
XM_011531465.2 |
3041 |
Missense Mutation |
AAA,AGA |
K868R |
XP_011529767.1 |
XM_017030066.1 |
3041 |
Missense Mutation |
AAA,AGA |
K927R |
XP_016885555.1 |
XM_017030067.1 |
3041 |
Missense Mutation |
AAA,AGA |
K897R |
XP_016885556.1 |
XM_017030068.1 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
XP_016885557.1 |
XM_017030069.1 |
3041 |
Missense Mutation |
AAA,AGA |
K852R |
XP_016885558.1 |
XM_017030070.1 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_016885559.1 |
XM_017030071.1 |
3041 |
Missense Mutation |
AAA,AGA |
K965R |
XP_016885560.1 |
XM_017030072.1 |
3041 |
Missense Mutation |
AAA,AGA |
K898R |
XP_016885561.1 |
XM_017030073.1 |
3041 |
Missense Mutation |
AAA,AGA |
K868R |
XP_016885562.1 |
XM_017030074.1 |
3041 |
Missense Mutation |
AAA,AGA |
K943R |
XP_016885563.1 |
View Full Product Details