Product Details

SNP ID
rs202150301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152168449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTTGAGGAGGCACTGGGAGCAG[C/T]GCCCTTGGAGATGGTGGAAAATTCA
Phenotype
MIM: 305660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRA3 PubMed Links

Gene Details

Gene
GABRA3
Gene Name
gamma-aminobutyric acid type A receptor alpha3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000808.3 1286 Missense Mutation ACT,GCT T420A NP_000799.1
XM_006724811.2 1286 UTR 3 XP_006724874.1

View Full Product Details