Product Details

SNP ID
rs200454840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78657759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGGGGCCTGTGGAAACTCTGAA[A/G]CTGCTGATGTCTCCACAACTATTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC5 PubMed Links

Gene Details

Gene
ZCCHC5
Gene Name
zinc finger CCHC-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152694.2 958 Missense Mutation GCT,GTT A221V NP_689907.1

View Full Product Details