Product Details

SNP ID
rs200689370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:11142256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTCATTCCTGGGTCTTTGGAT[C/T]CGCTTTTTAATGTTGAATGCCAAGT
Phenotype
MIM: 300118
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP6 PubMed Links

Gene Details

Gene
ARHGAP6
Gene Name
Rho GTPase activating protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287242.1 1884 Missense Mutation GAA,GGA E565G NP_001274171.1
NM_006125.2 1884 Intron NP_006116.2
NM_013423.2 1884 Missense Mutation GAA,GGA E542G NP_038267.1
NM_013427.2 1884 Missense Mutation GAA,GGA E745G NP_038286.2

View Full Product Details