Product Details

SNP ID
rs200819237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153803879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTCAGGGGGCCGTGGCCCAGC[C/T]CCTCCAGGCCTCGCGTCGGGGCCAG
Phenotype
MIM: 300089 MIM: 300634 MIM: 300090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IDH3G PubMed Links

Gene Details

Gene
IDH3G
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) gamma
There are no transcripts associated with this gene.

Gene
PDZD4
Gene Name
PDZ domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303512.1 2071 Missense Mutation GAG,GGG E601G NP_001290441.1
NM_001303513.1 2071 Missense Mutation GAG,GGG E499G NP_001290442.1
NM_001303514.1 2071 Missense Mutation GAG,GGG E486G NP_001290443.1
NM_001303515.1 2071 Missense Mutation GAG,GGG E520G NP_001290444.1
NM_001303516.1 2071 Missense Mutation GAG,GGG E514G NP_001290445.1
NM_032512.3 2071 Missense Mutation GAG,GGG E595G NP_115901.2
Gene
SSR4
Gene Name
signal sequence receptor subunit 4
There are no transcripts associated with this gene.

View Full Product Details