Product Details

SNP ID
rs201290923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37990950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTTCGTGGACCGCGTTGTGCA[A/G]GATGAACGAGACGTCCAAAGCCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYPM PubMed Links

Gene Details

Gene
HYPM
Gene Name
huntingtin interacting protein M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012274.1 134 Silent Mutation CAA,CAG Q37Q NP_036406.1
Gene
SYTL5
Gene Name
synaptotagmin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163334.1 134 Intron NP_001156806.1
NM_001163335.1 134 Intron NP_001156807.1
NM_138780.2 134 Intron NP_620135.1
XM_011544001.2 134 Intron XP_011542303.1
XM_011544002.2 134 Intron XP_011542304.1
XM_017029972.1 134 Intron XP_016885461.1
XM_017029973.1 134 Intron XP_016885462.1
XM_017029974.1 134 Intron XP_016885463.1

View Full Product Details