Product Details

SNP ID
rs201972199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:111776922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGACAGTTCAGTGCTGCTAGT[A/G]GAAAAGCTCCTTGGATGTTTTCTAT
Phenotype
MIM: 300334
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRPC5 PubMed Links

Gene Details

Gene
TRPC5
Gene Name
transient receptor potential cation channel subfamily C member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012471.2 3522 Silent Mutation TCC,TCT S771S NP_036603.1
XM_017029774.1 3522 Silent Mutation TCC,TCT S771S XP_016885263.1

View Full Product Details