Product Details

SNP ID
rs201092215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:64190636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGTTGAGGGCCATAGCAGCA[A/G]GTGGAGGTCGAGGGTGCAGGCCAGG
Phenotype
MIM: 300647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMER1 PubMed Links

Gene Details

Gene
AMER1
Gene Name
APC membrane recruitment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152424.3 2924 Missense Mutation CCT,CTT P884L NP_689637.3

View Full Product Details