Product Details

SNP ID
rs201642193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130402258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCCCCCGCCACCGTGAAGCCC[G/T]TTAACTAAGGTGAAGCTGATCAACG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RBMX2 PubMed Links

Gene Details

Gene
RBMX2
Gene Name
RNA binding motif protein, X-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016024.2 73 Silent Mutation NP_057108.2

View Full Product Details