Product Details

SNP ID
rs200754289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:139956051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTGTTCCTGGAATCTAACTTAC[A/G]AAAGGACCTGGAAGAGGACTTGACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf66 PubMed Links

Gene Details

Gene
CXorf66
Gene Name
chromosome X open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013403.2 953 Missense Mutation CGT,TGT R311C NP_001013421.1

View Full Product Details