Product Details

SNP ID
rs201993202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154295907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGTTCCCGGAGGAGGCCAGACC[C/T]GACAGGGGCACCTTGCAGGTGTTGC
Phenotype
MIM: 300092 MIM: 300044
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX28 PubMed Links

Gene Details

Gene
TEX28
Gene Name
testis expressed 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205201.1 234 Intron NP_001192130.1
NM_001586.2 234 Intron NP_001577.1
XM_011531116.1 234 Intron XP_011529418.1
XM_011531117.1 234 Intron XP_011529419.1
XM_011531118.2 234 Intron XP_011529420.1
Gene
TKTL1
Gene Name
transketolase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145933.1 234 Silent Mutation CCC,CCT P16P NP_001139405.1
NM_001145934.1 234 Intron NP_001139406.1
NM_012253.3 234 Silent Mutation CCC,CCT P16P NP_036385.3

View Full Product Details