Product Details

SNP ID
rs201151491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:69505943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCCGACAGCCTGGACTGTAGC[C/G]TGGACACCCTGATGGGGGACCTGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM155B PubMed Links

Gene Details

Gene
FAM155B
Gene Name
family with sequence similarity 155 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015686.2 709 Silent Mutation CTG,GTG L221V NP_056501.2
XM_011530908.2 709 Missense Mutation CTG,GTG L221V XP_011529210.1
XM_011530909.2 709 Missense Mutation CTG,GTG L221V XP_011529211.1

View Full Product Details