Product Details

SNP ID
rs201256036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:45151858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCCCCTGGAGAAGTCGAGGCA[A/G]CAACTTCTGACACACCAGCACCAGG
Phenotype
MIM: 300959
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf36 PubMed Links

Gene Details

Gene
CXorf36
Gene Name
chromosome X open reading frame 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024689.2 972 Intron NP_078965.2
NM_176819.3 972 Silent Mutation CTG,TTG L366L NP_789789.2
XM_005272670.1 972 Silent Mutation CTG,TTG L308L XP_005272727.1
XM_006724559.1 972 Silent Mutation CTG,TTG L282L XP_006724622.1

View Full Product Details