Product Details

SNP ID
rs2236207
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:56392509 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTTGACGCATTGGAGATAAAGTCC[A/G]AGGAGGTGAACTAACCTTTGAACGT
Phenotype
MIM: 603072 MIM: 600369
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKA PubMed Links

Gene Details

Gene
AURKA
Gene Name
aurora kinase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323303.1 139 Intron NP_001310232.1
NM_001323304.1 139 Intron NP_001310233.1
NM_001323305.1 139 Intron NP_001310234.1
NM_003600.3 139 Intron NP_003591.2
NM_198433.2 139 Intron NP_940835.1
NM_198434.2 139 Intron NP_940836.1
NM_198435.2 139 Intron NP_940837.1
NM_198436.2 139 Intron NP_940838.1
NM_198437.2 139 Intron NP_940839.1
XM_017028034.1 139 Intron XP_016883523.1
XM_017028035.1 139 Intron XP_016883524.1
Gene
CSTF1
Gene Name
cleavage stimulation factor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033521.1 139 UTR 5 NP_001028693.1
NM_001033522.1 139 Intron NP_001028694.1
NM_001324.2 139 Intron NP_001315.1
XM_011528600.1 139 Intron XP_011526902.1

View Full Product Details