Product Details
- SNP ID
-
rs5332
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:175441748 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGGATGAGCATGTGTGGCAGGATT[T/C]ATCTGCGAGTTCAGGTTGGGTGCTG
- Phenotype
-
MIM: 126449
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DRD1
PubMed Links
Gene Details
- Gene
- DRD1
- Gene Name
- dopamine receptor D1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000794.3 |
2297 |
UTR 3 |
|
|
NP_000785.1 |
View Full Product Details