Product Details

SNP ID
rs15612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:33988430 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGACCCACGGGGAAACAGGCCC[C/T]GAGTTACTGGATACAGGCAACCCTA
Phenotype
MIM: 604489 MIM: 606202
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMACR PubMed Links

Gene Details

Gene
AMACR
Gene Name
alpha-methylacyl-CoA racemase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167595.1 1747 Intron NP_001161067.1
NM_014324.5 1747 UTR 3 NP_055139.4
NM_203382.2 1747 UTR 3 NP_976316.1
Gene
C1QTNF3-AMACR
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SLC45A2
Gene Name
solute carrier family 45 member 2
There are no transcripts associated with this gene.

View Full Product Details