Product Details
- SNP ID
-
rs12587713
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:39265900 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGTAATACCTTATACCTTAACAATG[C/G]AGAATTACTAAGCAATTTTGGCTAC
- Phenotype
-
MIM: 602132
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CTAGE5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17109079] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTAGE5
- Gene Name
- CTAGE family member 5, ER export factor
- Gene
- LOC100288846
- Gene Name
- uncharacterized LOC100288846
There are no transcripts associated with this gene.
View Full Product Details