Product Details

SNP ID
rs2056420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16093646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCATTTTGGTTTCCAGAATATCAC[A/G]ATACCTAAACCACATCTAAACAAAT
Phenotype
MIM: 614631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISPD PubMed Links

Gene Details

Gene
ISPD
Gene Name
isoprenoid synthase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101417.3 Intron NP_001094887.1
NM_001101426.3 Intron NP_001094896.1
XM_006715770.3 Intron XP_006715833.1
XM_011515499.2 Intron XP_011513801.1
XM_011515500.2 Intron XP_011513802.1
XM_011515501.2 Intron XP_011513803.1
XM_011515508.2 Intron XP_011513810.1
XM_011515509.2 Intron XP_011513811.1
XM_017012575.1 Intron XP_016868064.1
XM_017012576.1 Intron XP_016868065.1
XM_017012577.1 Intron XP_016868066.1
XM_017012578.1 Intron XP_016868067.1

View Full Product Details