Product Details

SNP ID
rs12813582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122032828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCCCTCTTCATCTGTGAGCCAT[G/T]ATGTGCCATGTGCCACTCCCCCACC
Phenotype
MIM: 601406
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCL7A PubMed Links

Gene Details

Gene
BCL7A
Gene Name
BCL tumor suppressor 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024808.2 Intron NP_001019979.1
NM_020993.4 Intron NP_066273.1

View Full Product Details