Product Details
- SNP ID
-
rs28642812
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:123409363 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTATTGCCAGGCAAGGCCACGTG[C/T]GTGTCTTTCTGTTTTTACAATTTGA
- Phenotype
-
MIM: 607240
MIM: 614093
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KMT5A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138327565] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KMT5A
- Gene Name
- lysine methyltransferase 5A
- Gene
- RILPL2
- Gene Name
- Rab interacting lysosomal protein like 2
View Full Product Details