Product Details

SNP ID
rs34780788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121318067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTTACTGAATACACAGCTCA[A/G]GAAAGCCAGTGCCACTGGAAAGTGT
Phenotype
MIM: 606948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANAPC5 PubMed Links

Gene Details

Gene
ANAPC5
Gene Name
anaphase promoting complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137559.1 Intron NP_001131031.1
NM_016237.4 Intron NP_057321.2
XM_005253900.3 Intron XP_005253957.1
XM_017019423.1 Intron XP_016874912.1
XM_017019424.1 Intron XP_016874913.1

View Full Product Details