Product Details

SNP ID
rs11270
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156249580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGATATTCACCTCAACACTCC[A/T]AAAGCCAGCCCCTTCAGGTCTTCAG
Phenotype
MIM: 112260 MIM: 614579 MIM: 609176 MIM: 610962
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
There are no transcripts associated with this gene.

Gene
PMF1
Gene Name
polyamine-modulated factor 1
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323614.1 4153 UTR 3 NP_001310543.1
NM_001323615.1 4153 UTR 3 NP_001310544.1
NM_001323616.1 4153 UTR 3 NP_001310545.1
NM_001323617.1 4153 UTR 3 NP_001310546.1
NM_015327.2 4153 UTR 3 NP_056142.2
XM_017000842.1 4153 UTR 3 XP_016856331.1
XM_017000843.1 4153 UTR 3 XP_016856332.1

View Full Product Details