Product Details

SNP ID
rs10546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2973192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTCCCCACAGTCCGGGCCAGG[A/G]CAGCCTCAGGGGAGAGTGAAGGCCT
Phenotype
MIM: 609899 MIM: 614578 MIM: 602474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KREMEN2 PubMed Links

Gene Details

Gene
KREMEN2
Gene Name
kringle containing transmembrane protein 2
There are no transcripts associated with this gene.

Gene
PAQR4
Gene Name
progestin and adipoQ receptor family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284511.1 2295 UTR 3 NP_001271440.1
NM_001284512.1 2295 UTR 3 NP_001271441.1
NM_001284513.1 2295 UTR 3 NP_001271442.1
NM_001324118.1 2295 UTR 3 NP_001311047.1
NM_152341.4 2295 UTR 3 NP_689554.2
Gene
PKMYT1
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258450.1 2295 Missense Mutation GCC,GTC A376V NP_001245379.1
NM_001258451.1 2295 Missense Mutation GCC,GTC A436V NP_001245380.1
NM_004203.4 2295 Missense Mutation GCC,GTC A445V NP_004194.3
NM_182687.2 2295 Missense Mutation CCC,TCC P458S NP_872629.1
XM_011522734.2 2295 Missense Mutation GCC,GTC A445V XP_011521036.1
XM_011522735.2 2295 Missense Mutation GCC,GTC A436V XP_011521037.1
XM_011522736.2 2295 Missense Mutation GCC,GTC A436V XP_011521038.1

View Full Product Details