Product Details

SNP ID
rs11908486
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:37349700 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAAGCCCTTGCCTGGGTGATCTCAT[G/T]GAGTCTGAAGGTGCCGGTTGGGGCA
Phenotype
MIM: 190090
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SRC PubMed Links

Gene Details

Gene
SRC
Gene Name
SRC proto-oncogene, non-receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005417.4 Intron NP_005408.1
NM_198291.2 Intron NP_938033.1
XM_011529013.2 Intron XP_011527315.1
XM_017028024.1 Intron XP_016883513.1
XM_017028025.1 Intron XP_016883514.1
XM_017028026.1 Intron XP_016883515.1
XM_017028027.1 Intron XP_016883516.1

View Full Product Details