Product Details

SNP ID
rs11617888
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:45402214 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGATTAAAATTAAGTGATAGAACAA[A/G]TAAATTCAAAAAAGGAAAAAAGAAC
Phenotype
MIM: 610793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A30 PubMed Links

Gene Details

Gene
SLC25A30
Gene Name
solute carrier family 25 member 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010875.3 Intron NP_001010875.1
NM_001286806.1 Intron NP_001273735.1
NM_001286807.1 Intron NP_001273736.1
XM_005266321.2 Intron XP_005266378.1
XM_005266322.1 Intron XP_005266379.1
XM_006719793.3 Intron XP_006719856.1
XM_006719794.1 Intron XP_006719857.1
XM_011535026.1 Intron XP_011533328.1
XM_011535027.2 Intron XP_011533329.1
XM_017020523.1 Intron XP_016876012.1
XM_017020524.1 Intron XP_016876013.1

View Full Product Details