Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282447.2 | 2654 | UTR 3 | NP_001269376.1 | ||
NM_025220.4 | 2654 | UTR 3 | NP_079496.1 | ||
NM_153202.3 | 2654 | UTR 3 | NP_694882.1 | ||
XM_005260843.1 | 2654 | UTR 3 | XP_005260900.1 | ||
XM_006723639.1 | 2654 | UTR 3 | XP_006723702.1 | ||
XM_006723640.1 | 2654 | UTR 3 | XP_006723703.1 | ||
XM_006723644.2 | 2654 | Intron | XP_006723707.1 | ||
XM_011529366.1 | 2654 | UTR 3 | XP_011527668.1 | ||
XM_011529367.1 | 2654 | UTR 3 | XP_011527669.1 | ||
XM_011529368.1 | 2654 | UTR 3 | XP_011527670.1 | ||
XM_011529370.1 | 2654 | Intron | XP_011527672.1 | ||
XM_011529371.1 | 2654 | Intron | XP_011527673.1 | ||
XM_011529373.1 | 2654 | UTR 3 | XP_011527675.1 | ||
XM_017028080.1 | 2654 | Intron | XP_016883569.1 | ||
XM_017028081.1 | 2654 | Intron | XP_016883570.1 | ||
XM_017028082.1 | 2654 | Intron | XP_016883571.1 | ||
XM_017028083.1 | 2654 | Intron | XP_016883572.1 |