Product Details

SNP ID
rs677044
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:3668784 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTCTCCAGCCCTCAGGAACTTCTA[A/G]TGTGGCTCTGGGTTCCTGGAGTGGG
Phenotype
MIM: 607114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM33 PubMed Links

Gene Details

Gene
ADAM33
Gene Name
ADAM metallopeptidase domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282447.2 2686 UTR 3 NP_001269376.1
NM_025220.4 2686 UTR 3 NP_079496.1
NM_153202.3 2686 UTR 3 NP_694882.1
XM_005260843.1 2686 UTR 3 XP_005260900.1
XM_006723639.1 2686 UTR 3 XP_006723702.1
XM_006723640.1 2686 UTR 3 XP_006723703.1
XM_006723644.2 2686 Intron XP_006723707.1
XM_011529366.1 2686 UTR 3 XP_011527668.1
XM_011529367.1 2686 UTR 3 XP_011527669.1
XM_011529368.1 2686 UTR 3 XP_011527670.1
XM_011529370.1 2686 Intron XP_011527672.1
XM_011529371.1 2686 Intron XP_011527673.1
XM_011529373.1 2686 UTR 3 XP_011527675.1
XM_017028080.1 2686 Intron XP_016883569.1
XM_017028081.1 2686 Intron XP_016883570.1
XM_017028082.1 2686 Intron XP_016883571.1
XM_017028083.1 2686 Intron XP_016883572.1
Gene
GFRA4
Gene Name
GDNF family receptor alpha 4
There are no transcripts associated with this gene.

View Full Product Details