Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001732.3 | Intron | NP_001001732.2 | |||
NM_001159747.1 | Intron | NP_001153219.1 | |||
XM_011539789.2 | Intron | XP_011538091.1 | |||
XM_011539790.2 | Intron | XP_011538092.1 | |||
XM_011539791.2 | Intron | XP_011538093.1 | |||
XM_011539792.2 | Intron | XP_011538094.1 | |||
XM_011539793.2 | Intron | XP_011538095.1 | |||
XM_011539795.2 | Intron | XP_011538097.1 | |||
XM_017016240.1 | Intron | XP_016871729.1 |