Product Details

SNP ID
hCV11214086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46582131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCACAGCAGAAGTTATAGAGCGA[C/G]AACTTATATACACACCTAGAATGTA
Phenotype
MIM: 600153 MIM: 605857
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100506142 PubMed Links

Gene Details

Gene
LOC100506142
Gene Name
uncharacterized LOC100506142
There are no transcripts associated with this gene.

Gene
PIGF
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002643.3 1985 Intron NP_002634.1
NM_173074.2 1985 Intron NP_775097.1
Gene
RHOQ
Gene Name
ras homolog family member Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012249.3 1985 UTR 3 NP_036381.2

View Full Product Details