Product Details

SNP ID
rs34863464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113360428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATGGGGAGGGGGCTTTGGCCT[C/T]TGGAGGCCCCTCCTGGGCTGTGTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLBD2 PubMed Links

Gene Details

Gene
PLBD2
Gene Name
phospholipase B domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159727.1 Intron NP_001153199.1
NM_173542.3 Intron NP_775813.2
XM_011538023.2 Intron XP_011536325.1
XM_017018977.1 Intron XP_016874466.1

View Full Product Details