Product Details
- SNP ID
-
rs350107
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:69196754 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCTTTGTAGTGCCTTAATCCTTCCA[C/G]TGGCTTTGCACAAAAGTAGATGGTT
- Phenotype
-
MIM: 605607
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CENPH
PubMed Links
Gene Details
- Gene
- CENPH
- Gene Name
- centromere protein H
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022909.3 |
|
Intron |
|
|
NP_075060.1 |
View Full Product Details