Product Details

SNP ID
rs12540935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36153178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCACGTGCCCTCGGGAGTCCCG[C/G]GGAGGAGTTGCCGCGCGGAGGAGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EEPD1 PubMed Links

Gene Details

Gene
EEPD1
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030636.2 30 Intron NP_085139.2
XM_017012657.1 30 UTR 5 XP_016868146.1
XM_017012658.1 30 UTR 5 XP_016868147.1

View Full Product Details