Product Details

SNP ID
rs6963257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90405796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATAGGTAGAAGGAACTCTCAATG[G/T]TACCATCAAGAATTAAGTTAGGGCT
Phenotype
MIM: 611232
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLDN12 PubMed Links

Gene Details

Gene
CLDN12
Gene Name
claudin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185072.2 Intron NP_001172001.1
NM_001185073.2 Intron NP_001172002.1
NM_012129.4 Intron NP_036261.1
Gene
LOC101409256
Gene Name
cell division cycle 42 pseudogene
There are no transcripts associated with this gene.

Gene
LOC102723899
Gene Name
pituitary tumor-transforming gene 1 protein-interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011516764.2 Intron XP_011515066.1
XM_017012889.1 Intron XP_016868378.1
XM_017012890.1 Intron XP_016868379.1
XM_017012891.1 Intron XP_016868380.1
XM_017012892.1 Intron XP_016868381.1
XM_017012893.1 Intron XP_016868382.1
XM_017012894.1 Intron XP_016868383.1

View Full Product Details