Product Details

SNP ID
rs4735529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:98127382 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATATGTCATCCTGACATGATTA[C/T]CTTGTGTGCTACCTACTAGCTTTGT
Phenotype
MIM: 602486
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POP1 PubMed Links

Gene Details

Gene
POP1
Gene Name
POP1 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145860.1 Intron NP_001139332.1
NM_001145861.1 Intron NP_001139333.1
NM_015029.2 Intron NP_055844.2
XM_011516801.2 Intron XP_011515103.1

View Full Product Details