Product Details

SNP ID
rs7837618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86353245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTCTAACTGTTATTTAGGGCCA[A/C]AGTGATGACACCTACGGGCTACTTC
Phenotype
MIM: 602307
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WWP1 PubMed Links

Gene Details

Gene
WWP1
Gene Name
WW domain containing E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007013.3 Intron NP_008944.1
XM_005250760.3 Intron XP_005250817.1
XM_005250761.3 Intron XP_005250818.1
XM_011516802.2 Intron XP_011515104.1
XM_017012993.1 Intron XP_016868482.1
XM_017012994.1 Intron XP_016868483.1
XM_017012995.1 Intron XP_016868484.1
XM_017012996.1 Intron XP_016868485.1
XM_017012997.1 Intron XP_016868486.1

View Full Product Details