Product Details

SNP ID
rs12406968
Assay Type
Validated
NCBI dbSNP Submissions
42
Location
Chr.1:28745278 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTAGGGATGGTTGCTATTTTATCTT[C/T]CAGTGGGTACAGATTAATAAGAAAT
Phenotype
MIM: 610640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YTHDF2 PubMed Links

Gene Details

Gene
YTHDF2
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172828.1 Intron NP_001166299.1
NM_001173128.1 Intron NP_001166599.1
NM_016258.2 Intron NP_057342.2

View Full Product Details