Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142641.1 | Intron | NP_001136113.1 | |||
XM_005266171.3 | Intron | XP_005266228.1 | |||
XM_005266173.3 | Intron | XP_005266230.1 | |||
XM_005266175.3 | Intron | XP_005266232.1 | |||
XM_005266176.3 | Intron | XP_005266233.1 | |||
XM_005266177.3 | Intron | XP_005266234.1 | |||
XM_005266181.3 | Intron | XP_005266238.1 | |||
XM_011534803.2 | Intron | XP_011533105.1 | |||
XM_011534804.2 | Intron | XP_011533106.1 | |||
XM_011534805.2 | Intron | XP_011533107.1 | |||
XM_011534806.2 | Intron | XP_011533108.1 | |||
XM_011534807.2 | Intron | XP_011533109.1 | |||
XM_011534808.2 | Intron | XP_011533110.1 | |||
XM_011534809.2 | Intron | XP_011533111.1 | |||
XM_011534810.2 | Intron | XP_011533112.1 | |||
XM_011534812.2 | Intron | XP_011533114.1 | |||
XM_011534816.2 | Intron | XP_011533118.1 |