Product Details

SNP ID
rs2500018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5046947 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTACGCAACCATCTTGACATCCC[A/T]AGTGTTGGTGGGCATTAGCATGTGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52J3 PubMed Links

Gene Details

Gene
OR52J3
Gene Name
olfactory receptor family 52 subfamily J member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001916.2 422 Missense Mutation CAA,CTA Q141L NP_001001916.2

View Full Product Details