Product Details

SNP ID
rs9932581
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88651945 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAACGGCGGAGGCCGGAGGCAG[C/T]AATGCTGGTTTATTCCCCATGGCCA
Phenotype
MIM: 608508 MIM: 603236
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CYBA PubMed Links

Gene Details

Gene
CYBA
Gene Name
cytochrome b-245 alpha chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000101.3 1958 Intron NP_000092.2
XM_011522905.2 1958 Intron XP_011521207.1
Gene
MVD
Gene Name
mevalonate diphosphate decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002461.2 1958 UTR 3 NP_002452.1
XM_011523086.2 1958 UTR 3 XP_011521388.1
XM_011523087.2 1958 UTR 3 XP_011521389.1
XM_011523088.2 1958 UTR 3 XP_011521390.1
XM_011523089.2 1958 UTR 3 XP_011521391.1

View Full Product Details