Product Details
- SNP ID
-
rs2673051
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:45690917 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGTTTAATGTGATCCTACACCAG[G/T]TCTCAAATGTGTTTAAGCCATATGG
- Phenotype
-
MIM: 604543
MIM: 606569
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LIMD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142719743] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LIMD1
- Gene Name
- LIM domains containing 1
There are no transcripts associated with this gene.
- Gene
- LIMD1-AS1
- Gene Name
- LIMD1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- SACM1L
- Gene Name
- SAC1 suppressor of actin mutations 1-like (yeast)
View Full Product Details